Hexamethyleneimine, 99%
Hexamethyleneimine, 99%
Hexamethyleneimine, 99%
Acros Organics

Hexamethyleneimine, 99%

CAS: 111-49-9 | C6H13N | 99.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
120595000
also known as 12059
500 mLGlass bottle
Catalog number 120595000
also known as 12059
HSN 29252990
Price (INR)
11,741.00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS111-49-9
IUPAC Nameazepane
Molecular FormulaC6H13N
InChI KeyZSIQJIWKELUFRJ-UHFFFAOYSA-N
SMILESC1CCCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4650 to 1.4670 (20°C, 589 nm)
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only