5-Hydroxyindole, 97%
5-Hydroxyindole, 97%
5-Hydroxyindole, 97%
Acros Organics

5-Hydroxyindole, 97%

CAS: 1953-54-4 | C8H7NO | 133.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
5 g
Packaging:
Glass bottle
Catalog number 121270010
HSN 29339900
Price (INR)
9,871.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,871.00
Each
Chemical Identifiers
CAS1953-54-4
IUPAC Name1H-indol-5-ol
Molecular FormulaC8H7NO
InChI KeyLMIQERWZRIFWNZ-UHFFFAOYSA-N
SMILESOC1=CC=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point105°C to 110°C
Appearance (Form)Crystalline powder or needles
Appearance (Color)Beige to brown
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only