Insulation Rings
Actual product may vary
Thermo Scientific™
Insulation Rings
Choose quality consumables such as these insulation rings to ensure consistent, reliable performance from the Thermo Scientific™ ELEMENT GD™ GD-MS system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1212920Isolation Ring Ceramic
1249330Isolation Ring Sapphire
Catalog number 1212920
Price (INR)
-
Type:
Isolation Ring Ceramic
Choose quality consumables such as these Thermo Scientific™ Insulation Rings to ensure consistent, reliable performance from the Thermo Scientific™ ELEMENT™ GD GD-MS system. Due to the inevitable deterioration of insulation materials as a result of the glow discharge process, regular replacement of the insulation rings is essential.
Ensure Peak Performance with Quality Consumables
  • Sapphire insulation ring, resistant to cleaning with hydrofluoric acid
  • Ceramic insulation ring, not resistant to cleaning with hydrofluoric acid
Specifications
For Use With (Equipment)ELEMENT GD GD-MS Instrument
TypeIsolation Ring Ceramic
Unit SizeEach