2-Hydroxy-p-naphthoquinone, 99%
2-Hydroxy-p-naphthoquinone, 99%
2-Hydroxy-p-naphthoquinone, 99%
Acros Organics

2-Hydroxy-p-naphthoquinone, 99%

CAS: 83-72-7 | C10H6O3 | 174.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
12163010010 gGlass bottle
Catalog number 121630100
HSN 29146990
Price (INR)
6,060.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
6,060.00
Each
Chemical Identifiers
CAS83-72-7
IUPAC Name4-hydroxy-1,2-dihydronaphthalene-1,2-dione
Molecular FormulaC10H6O3
InChI KeyWVCHIGAIXREVNS-UHFFFAOYSA-N
SMILESOC1=CC(=O)C(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=98.5 %
Appearance (Color)Yellow to orange to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only