Allopurinol, 98%
Allopurinol, 98%
Allopurinol, 98%
Acros Organics

Allopurinol, 98%

CAS: 315-30-0 | C5H4N4O | 136.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 121830050
HSN 29335990
Price (INR)
3,787.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
3,787.00
Each
Chemical Identifiers
CAS315-30-0
IUPAC Name1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
Molecular FormulaC5H4N4O
InChI KeyOFCNXPDARWKPPY-UHFFFAOYSA-N
SMILESO=C1N=CN=C2NNC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to almost white
Loss on drying=<0.5 % (1 g, 105°C)
Appearance (Form)Powder
Titration with NaOH>=97.5 % (on dried substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only