Product Image
Invitrogen™

PureLink™ 96 Receiver Plates

PureLink™ 96: Sample size 1.3 ml culture; Typical yields up to 3-5 μgPureLink™ 384: Sample size 200 μl culture; TypicalRead more
Have Questions?
Catalog NumberQuantity
1219302550 Plates
Catalog number 12193025
Price (INR)
88,497.00
Each
Quantity:
50 Plates
Price (INR)
88,497.00
Each
PureLink™ 96: Sample size 1.3 ml culture; Typical yields up to 3-5 μg
PureLink™ 384: Sample size 200 μl culture; Typical yields up to 2-3 μg

––Purify 96 or 384 plasmid DNA samples simultaneously
–centrifugation method eliminates cross-contamination


The PureLink™ 96 Plasmid Purification System is designed for high-throughput isolation of high-copy number plasmids. Following the PureLink™ 96 Plasmid Purification Systems protocol, up to 96 plasmid DNA samples can be simultaneously purified. The novel technology eliminates time-consuming cell harvesting and alkaline lysis steps, saving time and effort. The unique centrifugationbased format of the PureLink™ 96 Plasmid Purification System eliminates opportunities for cross-contamination between wells, a critical factor in PCR and microarrays. The isolated plasmid DNA is ideal for use in automated sequencing reactions, offering superior read lengths and data quality. Samples may be processed 96 at a time or in smaller increments. The PureLink™ 96 Plasmid Purification System is compatible with a variety of centrifuges, robots, pipetting devices, automated sequencers, and sequencing chemistries.

The PureLink™ 384 Plasmid Purification Plate is designed for ultra highthroughput DNA isolation. By utilizing the same technology as the PureLink™ 96 Plasmid Purification System, the PureLink™ 384 Plasmid Purification Plate reduces time and effort while reliably isolating high-quality plasmid DNA for DNA sequencing, PCR, and restriction digestion.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Automated Sequencing Reaction
For Use With (Equipment)Robotic Liquid Handling Systems
Plate TypeReceiver Plate
Product LinePureLink
Product TypeReceiver Plate
Quantity50 Plates
Shipping ConditionRoom Temperature
Format96-well Plate
Unit SizeEach
Contents & Storage
The PureLink™ 96 Plasmid Purification System is available as a complete kit and includes filter plates, receiver plates, air porous tape, foil tape, growth blocks, lysis buffer, TE buffer, cell suspension buffer, and RNase A. Components are also available separately. PureLink™ 384 Plasmid Purification Plates are available in packs of 25. PureLink™ 384 Pre-Lysis Buffer is also available separately. Store at room temperature. All components are guaranteed stable for 2 years when properly stored.