The PureLink™ Growth Blocks are a component in the PureLink™ 96 Plasmid Purification System. The PureLink™ 96 Plasmid Purification SystemRead more
Have Questions?
Catalog Number
Quantity
12256020
also known as 12256-020
50 Blocks
Catalog number 12256020
also known as 12256-020
Price (INR)
92,523.00
Each
Quantity:
50 Blocks
Price (INR)
92,523.00
Each
The PureLink™ Growth Blocks are a component in the PureLink™ 96 Plasmid Purification System. The PureLink™ 96 Plasmid Purification System offers a unique approach to high throughput plasmid DNA isolation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Automated Nucleic Acid Purification, DNA Extraction
Product LinePureLink
Product TypeGrowth Block
Quantity50 Blocks
Shipping ConditionRoom Temperature
Format96 Deep-well Plate
Unit SizeEach
Contents & Storage
Package of 50 blocks The PureLink™ 96 Components are shipped at room temperature. Upon receipt, store the components at room temperature.