One Shot&trade; MAX Efficiency&trade; DH5&alpha;-T1<sup>R</sup> Competent Cells
Invitrogen™

One Shot™ MAX Efficiency™ DH5α-T1R Competent Cells

One Shot MAX Efficiency DH5α-T1R Competent Cells are derived from the popular DH5α strain and carry the tonA (fhuA) mutationRead more
Have Questions?
Catalog NumberQuantity
1229701621 x 50 μL
Catalog number 12297016
Price (INR)
55,819.00
Each
Quantity:
21 x 50 μL
Price (INR)
55,819.00
Each

One Shot MAX Efficiency DH5α-T1R Competent Cells are derived from the popular DH5α strain and carry the tonA (fhuA) mutation that confers resistance to T1 and T5 phage. They offer added security to protect valuable clones and libraries. This is especially important for genome and sequencing centers where T1 phage infection would be detrimental.

DH5α-T1R is a versatile strain that can be used in many everyday cloning applications. Prepared by a proprietary modification of the Hanahan method, these cells are suitable for the construction of gene banks or for the generation of cDNA libraries using plasmid-derived vectors. The lacZ∆M15 marker provides α-complementation of the ß-galactosidase gene from pUC or similar vectors and therefore can be used for blue/white screening of colonies on bacterial plates containing Bluo-gal or X-gal. DH5α-T1R Competent Cells are capable of being transformed efficiently with large plasmids and can also serve as a host for the M13mp cloning vectors if a lawn of DH5αF'IQ is provided to allow plaque formation.

Benefits of MAX Efficiency DH5α-T1R Competent Cells
• Transformation efficiency of >1 x 109 transformants/μg with non-saturating amounts of DNA
tonA (fhuA) mutation to confer resistance to T1 and T5 phage
endA1 mutation that enables cleaner DNA preparations and better results in downstream applications due to the elimination of nonspecific digestion by Endonuclease I
recA1 mutation that reduces occurrence of unwanted recombination in cloned DNA
hsdR for efficient transformation of unmethylated DNA from PCR applications  
lacZΔM15 marker that allows blue/white color screening of recombinant clones carrying the alpha fragment of β-galactosidase 
 
Convenient and efficient format 
DH5α-T1R E. coli are supplied in the convenient, single-reaction One Shot format. Each tube contains enough cells for one transformation so there are no efficiency-zapping, freeze-thaw cycles or money wasted on unused cells.

Genotype: F- φ80lacZΔM15 Δ(lacZYA-argF)U169 recA1 endA1 hsdR17(rK-, mK+) phoA supE44 thi-1 gyrA96 relA1 tonA

Find the strain and format that fits you needs
DH strains are available in chemically competent and electrocompetent cell formats.
MAX Efficiency DH5αF´IQ Competent Cells are available in chemically competent format.
DH5α-T1R is available in Multishot format for high throughput applications.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Antibiotic Resistance BacterialNo
Blue/White ScreeningYes (lacZΔM15)
Cloning Methylated DNANo
Cloning Unstable DNANot suitable for cloning unstable DNA
Contains F' EpisomeNo
High-throughput CompatibilityLow
Improves Plasmid QualityYes (endA1)
Preparing Unmethylated DNANo
Product LineOne Shot
Product TypeChemically Competent Cells
Quantity21 x 50 μL
Reduces RecombinationYes (recA1)
Shipping ConditionDry Ice
T1 Phage - Resistant (tonA)Yes
Transformation Efficiency LevelHigh Efficiency (>1 x 109 cfu/μg)
FormatTube
SpeciesE. coli (K12)
Unit SizeEach
Contents & Storage
• One Shot MAX Efficiency DH5α-T1R Competent Cells (21 x 50 μL)
Store Competent Cells at –80°C.

• pUC19 DNA (50 μL 10 pg/μL)
Store pUC19 DNA at –20°C.

• S.O.C. Medium (6 mL)
Store S.O.C. Medium at 4°C or room temperature.

Frequently asked questions (FAQs)

How do you recommend that I prepare my DNA for successful electroporation of E. coli?

For best results, DNA used in electroporation must have a very low ionic strength and a high resistance. A high-salt DNA sample may be purified by either ethanol precipitation or dialysis.

The following suggested protocols are for ligation reactions of 20ul. The volumes may be adjusted to suit the amount being prepared.

Purifying DNA by Precipitation: Add 5 to 10 ug of tRNA to a 20ul ligation reaction. Adjust the solution to 2.5 M in ammonium acetate using a 7.5 M ammonium acetate stock solution. Mix well. Add two volumes of 100 % ethanol. Centrifuge at 12,000 x g for 15 min at 4C. Remove the supernatant with a micropipet. Wash the pellet with 60ul of 70% ethanol. Centrifuge at 12,000 x g for 15 min at room temperature. Remove the supernatant with a micropipet. Air dry the pellet. Resuspend the DNA in 0.5X TE buffer [5 mM Tris-HCl, 0.5 mM EDTA (pH 7.5)] to a concentration of 10 ng/ul of DNA. Use 1 ul per transformation of 20 ul of cell suspension.

Purifying DNA by Microdialysis: Float a Millipore filter, type VS 0.025 um, on a pool of 0.5X TE buffer (or 10% glycerol) in a small plastic container. Place 20ul of the DNA solution as a drop on top of the filter. Incubate at room temperature for several hours. Withdraw the DNA drop from the filter and place it in a polypropylene microcentrifuge tube. Use 1ul of this DNA for each electrotransformation reaction.

You offer competent cells in Subcloning Efficiency, Library Efficiency and MAX Efficiency. How do these differ?

There are a few exceptions, but in general the difference is in guaranteed transformation efficiency as follows:

Subcloning Efficiency cells are guaranteed to produce at least 1.0 x 10E6 transformants per µg of transformed pUC19 or pUC18 supercoiled plasmid
Library Efficiency cells are guaranteed to produce at least 1.0 x 10E8 transformants per µg pUC19 or pUC18 DNA
MAX Efficiency cells are guaranteed to produce at least 1.0 x 10E9 transformants per µg pUC19 or pUC18 DNA

Do any Invitrogen competent cells contain DMSO in the freezing medium?

Yes, several of our competent cells products are frozen with DMSO. The presence of DMSO (dimethylsulfoxide) will generally be indicated in the MSDS files if you have a question about a particular product, but here is a list of commonly used products that are known to have DMSO in the freezing buffer:

One Shot OmniMAX 2 T1 Phage Resistant Cells, Cat. No. C8540-03

One Shot INV?F' Chemically Competent Cells, Cat. No. C2020-03 and C2020-06

One Shot MAX Efficiency DH5?-T1 Chemically Competent Cells, Cat. No. 12297-016

MAX Efficiency DH5?-T1 Phage Resistant Cells, Cat. No. 12034-013

MAX Efficiency DH5? Chemically Competent Cells, Cat. No. 18258-012

Library Efficiency DH5? Chemically Competent Cells, Cat. No. 18263-012

MAX Efficiency DH5? F'IQ Cells, Cat. No. 18288-019

MAX Efficiency Stbl2Chemically Competent Cells, Cat. No. 10268-019

When should DMSO, formamide, glycerol and other cosolvents be used in PCR?

Cosolvents may be used when there is a failure of amplification, either because the template contains stable hairpin-loops or the region of amplification is GC-rich. Keep in mind that all of these cosolvents have the effect of lowering enzyme activity, which will decrease amplification yield. For more information see P Landre et al (1995). The use of co-solvents to enhance amplification by the polymerase chain reaction. In: PCR Strategies, edited by MA Innis, DH Gelfand, JJ Sninsky. Academic Press, San Diego, CA, pp. 3-16.

Additionally, when amplifying very long PCR fragments (greater than 5 kb) the use of cosolvents is often recommended to help compensate for the increased melting temperature of these fragments.

Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.

Citations & References (1)

Citations & References
Abstract
Haemophilus influenzae type b strain A2 has multiple sialyltransferases involved in lipooligosaccharide sialylation.
Authors: Jones Paul A; Samuels Nicole M; Phillips Nancy J; Munson Robert S Jr; Bozue Joel A; Arseneau Julie A; Nichols Wade A; Zaleski Anthony; Gibson Bradford W; Apicella Michael A;
Journal:J Biol Chem
PubMed ID:11842084
The lipooligosaccharide (LOS) of Haemophilus influenzae contains sialylated glycoforms, and a sialyltransferase, Lic3A, has been previously identified. We report evidence for two additional sialyltransferases, SiaA, and LsgB, that affect N-acetyllactosamine containing glycoforms. Mutations in genes we have designated siaA and lsgB affected only the sialylated glycoforms containing N-acetylhexosamine. A mutation ... More