Torch Holder for iCAP™ Q/Qnova/MX Series ICP-MS
Thermo Scientific™
Torch Holder for iCAP™ Q/Qnova/MX Series ICP-MS
Elevate ICP‑MS performance with this Thermo Scientific™ Torch Holder for ICP‑MS instruments, which locks alignment, boosts stability, and increases uptime. Reduce drift, protect the torch, and keep your lab running with consistent, repeatable results—every sample, every time.
Have Questions?
Catalog NumberFor Use With (Equipment)
1231000iCAP MX Series ICP-MS, iCAP Q/Qnova ICP-MS
Catalog number 1231000
Price (INR)
-
For Use With (Equipment):
iCAP MX Series ICP-MS, iCAP Q/Qnova ICP-MS
Boost accuracy and uptime with this Torch Holder for ICP‑MS instruments, engineered to lock torch alignment for consistent, repeatable results. The rigid, corrosion‑resistant frame minimizes drift and vibration, protects fragile components, and accelerates torch maintenance without tools. Precision guides and a positive‑stop mechanism ensure quick, error‑free installation, while high‑temperature materials withstand daily use. Streamline workflows, cut re‑optimization time, and keep your ICP‑MS system running at peak performance.
Improve alignment consistency
  • Maintain coaxial geometry across runs and prevent mis-seating during maintenance for stable, repeatable plasma coupling.
Reduce re-optimization time
  • Enable rapid torch swaps without tools
  • Preserve previous tuning parameters
Specifications
For Use With (Equipment)iCAP MX Series ICP-MS, iCAP Q/Qnova ICP-MS
TypeTorch Holder
Unit SizeEach