Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid
Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid
Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid
Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid
Acros Organics

Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid

Hydrogen bromide, 31 to 34 wt% (HBr), BrH, CAS Number-10035-10-6, 64-19-7, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, Vapor Pressure: 300mmHg at 20 deg.C | CAS: 10035-10-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
View all
Glass bottle
Catalog number 123180025
also known as 12318
HSN 28111990
Price (INR)
40,502.00
Each
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
40,502.00
Each
Add to cart
Chemical Identifiers
CAS37348-16-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration with NaOH31.5 to 36.5 % (HBr)
Water=<0.5 % (K.F.)
Appearance (Color)Clear light yellow to orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only