Hesperidin, 90+%
Hesperidin, 90+%
Hesperidin, 90+%
Acros Organics

Hesperidin, 90+%

CAS: 520-26-3 | C28H34O15 | 610.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
Packaging:
View all
Glass Bottle
Catalog number 123460050
HSN 29389090
Price (INR)
3,016.00
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS520-26-3
IUPAC Name(2S)-5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}-3,4-dihydro-2H-1-benzopyran-4-one
Molecular FormulaC28H34O15
InChI KeyQUQPHWDTPGMPEX-QJBIFVCTSA-N
SMILESCOC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2O1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=90.0 %
Infrared spectrumConforms
Loss on drying=<5 %
Residue after ign. (sulfated)=<0.3 %
Heavy metals=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only