Platinum™ SuperFi™ PCR Master Mix
Invitrogen™

Platinum™ SuperFi™ PCR Master Mix

Invitrogen Platinum SuperFi PCR Master Mix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
12358010100 Reactions
12358050500 Reactions
123582505 x 500 Reactions
Catalog number 12358010
Price (INR)
26,054.00
Online offer
Ends: 31-Dec-2026
37,219.00
Save 11,165.00 (30%)
Each
No. of Reactions:
100 Reactions
Request bulk or custom format
Price (INR)
26,054.00
Online offer
Ends: 31-Dec-2026
37,219.00
Save 11,165.00 (30%)
Each
Invitrogen Platinum SuperFi PCR Master Mix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. Only template, primers, and water need to be added, thus reducing the number of pipetting steps during PCR reaction setup.

Platinum SuperFi PCR Master Mix retains all the features of Platinum SuperFi DNA Polymerase, a proofreading DNA polymerase that combines superior fidelity with trusted Platinum hot-start technology. Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.

Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation.

Features

  • Exceptional >300X Taq fidelity
  • High specificity and increased yields with Platinum hot-start technology
  • Robust amplification of difficult targets of high GC content
  • Room temperature reaction setup and 24-hour bench top stability of pre-assembled reactions
  • Amplification of samples with suboptimal purity

Applications

  • High-fidelity PCR
  • Cloning and sub-cloning
  • Site-directed mutagenesis
  • Amplification of GC-rich templates
  • Template generation for sequencing
  • High-throughput PCR
  • Long PCR

Notes

  • Annealing rules for Platinum SuperFi DNA Polymerase are different from many common DNA polymerases.
  • Platinum SuperFi PCR Master Mix is supplied with separate SuperFi GC Enhancer for difficult and GC-rich templates (˃65% GC).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)300X
Hot StartBuilt-In Hot Start
No. of Reactions100 Reactions
OverhangBlunt
PolymerasePlatinum SuperFi DNA Polymerase
Quantity100 reactions
Reaction FormatSuperMix or Master Mix
Shipping ConditionWet or Dry Ice
Size (Final Product)20 kb or less
For Use With (Application)Hot-start PCR, High-fidelity PCR
GC-Rich PCR PerformanceHigh
Reaction SpeedFast
Unit SizeEach
Contents & Storage
• 2X Platinum SuperFi PCR Master Mix (2 x 1.25 mL)
• SuperFi GC Enhancer (1.25 mL)
• Nuclease-free water (2 x 1.25 mL)

Store at –5°C to –30°C.

Frequently asked questions (FAQs)

Which nucleotide analogues can be used with Platinum SuperFi DNA Polymerase?

Platinum SuperFi DNA Polymerase cannot read dUTP-derivatives or dITP in DNA templates, so the use of these analogues is not recommended.Platinum SuperFi DNA Polymerase can incorporate 7-deaza-dGTP and radiolabeled dNTPs. 

I am having difficulties amplifying long targets using Platinum SuperFi DNA Polymerase. What are your recommendations?

Good lab practices are important for long fragment amplification. These include using high-quality templates (pure, fresh and intact) and fresh primer solutions. Optimization steps to consider include decreasing denaturation and extension temperatures, lengthening extension times as recommended in the manual, and increasing template amounts.

Can Platinum SuperFi DNA Polymerase amplify AT-rich targets?

Yes. To improve amplification of AT-rich targets, we recommend reducing the extension temperature to 68 degrees C or add 5-15 mM Tetramethylammonium Chloride (TMAC).

Can Platinum SuperFi DNA Polymerase amplify GC-rich targets?

All Platinum SuperFi product formats are supplied with 5X SuperFi GC Enhancer which is optimized to improve amplification of GC-rich targets (recommended for use with targets containing >65% GC content)

Does SuperFi Green Buffer influence performance of Platinum SuperFi DNA Polymerase?

No. The colored buffer does not interfere with PCR performance and does not change enzyme features.