Sodium carbonate, 99.5%, extra pure, anhydrous
Sodium carbonate, 99.5%, extra pure, anhydrous
Sodium carbonate, 99.5%, extra pure, anhydrous
Sodium carbonate, 99.5%, extra pure, anhydrous
Acros Organics

Sodium carbonate, 99.5%, extra pure, anhydrous

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
1 kg
2.5 kg
Packaging:
View all
Plastic Drum
Catalog number 123670100
HSN 28362090
Price (INR)
65,086.00
Each
Quantity:
10 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (INR)
65,086.00
Each
Chemical Identifiers
CAS497-19-8
IUPAC Namedisodium carbonate
Molecular FormulaCNa2O3
InChI KeyCDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<1 % (300°C, 2 h)

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Adsorbent
RUO – Research Use Only