ESI DX FAST Autosamplers
Actual product may vary
ESI DX FAST Autosamplers
ESI DX FAST Autosamplers
ESI DX FAST Autosamplers
ESI DX FAST Autosamplers
Thermo Scientific™
ESI DX FAST Autosamplers
Increase productive instrument time by reducing sample uptake, stabilization, and rinse steps while maintaining superior analytical results with ESI DX Autsamplers with SC-FAST systems
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
1238550ESI SC-14 DX Autosampler FASTHR-ICP-MS System
1329540ESI SC-2 DX Autosampler FASTICP-OES System, ICP-MS Systems
1329550ESI SC-4 DX Autosampler FASTICP-MS Systems
1329560ESI SC-8 DX Autosampler FASTICP-OES System, ICP-MS Systems
1329570ESI SC-14 DX Autosampler FASTICP-OES System, ICP-MS Systems
842347004007ESI SC-4 DX Autosampler FASTICP-OES System
BRE0014202ESI SC-4 DX Autosampler FASTHR-ICP-MS System
BRE0014294ESI SC-2 DX Autosampler FASTHR-ICP-MS Systems
Catalog number 1238550
Price (INR)
-
Type:
ESI SC-14 DX Autosampler FAST
For Use With (Equipment):
HR-ICP-MS System
Experience automated high-throughput sample introduction into your ICP-OES and ICP-MS systems and improve sample throughput 2–5x over standard autosamplers with Elemental Scientific™ (ESI) SC-DX FAST autosampler systems. These systems use valve injection with rapid vacuum sample loading to increase productive instrument time by reducing sample uptake, stabilization, and rinse steps while maintaining superior analytical results. ESI DX FAST systems also reduce laboratory operating costs by decreasing consumption of argon, power, and reagents.
ESI DX FAST autosampler systems provide high-throughput, automated sample introduction for Thermo Scientific™ ICP-OES and ICP-MS systems. These powerful autosamplers are controlled by fully integrated software plugins within Thermo Scientific™ Qtegra™ Intelligent Scientific Data Solution™ (ISDS) or ESI Software.
Compatible with:
  • Thermo Scientific™ iCAP™ 6000/7000/PRO Series ICP-OES Systems
  • Thermo Scientific™ iCAP™ Q/RQ/TQ Series ICP-MS Systems
  • Thermo Scientific™ Element Series HR-ICP-MS Systems (SC-2DX, SC-4DX and SC-14DX only) with ESI SC software
ESI DX FAST systems offer:
  • Rapid vacuum loading and valve injection for 2–5x greater sample throughput than standard peristaltic pump uptake
  • Enhanced washout from specifically designed P-Series valves
  • Reduced laboratory costs for argon, power, and reagents through reduced analysis times
  • Improved efficiency from minimal instrument exposure to sample matrix
ESI DX FAST Autosampler system models
  • 2DX FAST Autosampler—2 large racks, 4 microplates
  • 4DX FAST Autosampler—4 large racks, 6 microplates
  • 8DX FAST Autosampler—8 large racks, supplied with Mobile Station
  • 14DX FAST Autosampler—14 large racks, supplied with Mobile Station
Specifications
For Use With (Equipment)HR-ICP-MS System
TypeESI SC-14 DX Autosampler FAST
Unit SizeEach