Product Image
Gibco™

Ham's F-10 Nutrient Mix, HEPES

Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells. F-10 has since beenRead more
Have Questions?
Catalog NumberQuantity
12390035500 mL
Catalog number 12390035
Price (INR)
3,156.00
Online offer
Ends: 31-Dec-2026
5,259.00
Save 2,103.00 (40%)
Each
Quantity:
500 mL
Customize this product
Price (INR)
3,156.00
Online offer
Ends: 31-Dec-2026
5,259.00
Save 2,103.00 (40%)
Each

Ham's F-10 Nutrient Mixture (F-10) was designed for serum-free growth of Chinese Hamster Ovary (CHO) cells. F-10 has since been used for serum-free growth of CHO cultures as well as serum-supplemented growth of other mammalian cells, including COS-7, primary rat astrocytes, and rat prostate epithelial cells.


This Ham's F-10 Nutrient Mixture is modified as follows:
With
• Phenol Red
• HEPES
• L-glutamine


The complete formulation is available.

Compared to other basal media, F-10 contains a wider variety of components, including zinc, hypoxanthine, and thymidine. Serum-free growth of CHO cells in F-10 has led to a variety of improved formulations, such as Ham's F-12 Nutrient Mixture (F-12).

Using Ham's F-10 Nutrient Mixture
Gibco™ F-10 contains no proteins or growth factors. Therefore, F-10 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). F-10 uses a sodium bicarbonate buffer system (1.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
 

Specifications
Cell LineCHO, COS-7, and rat prostate epithelial cells
Cell TypePrimary Rat Astrocytes
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeHam's F-10 Nutrient Mixture
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
SterilitySterile-filtered
With AdditivesHigh Glucose, Glutamine, HEPES, Phenol Red, Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I see a decrease in growth of my culture. What should I do?

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

My cells are not adhering to the culture vessel. What should I do?

This can occur if cells are overly trypsinized. Trypsinize for a shorter time or use less trypsin. Mycoplasma contamination could also cause this problem. Segregate your culture and test for mycoplasma infection. Lastly, check for attachment factors in the medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (2)

Citations & References
Abstract
The carboxyl terminus of GLUT4 contains a serine-leucine-leucine sequence that functions as a potent internalization motif in Chinese hamster ovary cells.
Authors: Garippa R J; Johnson A; Park J; Petrush R L; McGraw T E;
Journal:J Biol Chem
PubMed ID:8702815
'To characterize the trafficking motifs contained in the carboxyl terminus of GLUT4, a chimera (GTCTR) was constructed in which the carboxyl-terminal 30 amino acids of GLUT4 were substituted for the amino-terminal cytoplasmic domain of the transferrin receptor (TR). The endocytic behavior of this chimera was characterized in Chinese hamster ovary ... More
Subretinal transplantation of genetically modified human cell lines attenuates loss of visual function in dystrophic rats.
Authors: Lund R D; Adamson P; Sauvé Y; Keegan D J; Girman S V; Wang S; Winton H; Kanuga N; Kwan A S; Beauchène L; Zerbib A; Hetherington L; Couraud P O; Coffey P; Greenwood J;
Journal:Proc Natl Acad Sci U S A
PubMed ID:11504951
'Royal College of Surgeons rats are genetically predisposed to undergo significant visual loss caused by a primary dysfunction of retinal pigment epithelial (RPE) cells. By using this model, we have examined the efficacy of subretinal transplantation of two independent human RPE cell lines each exhibiting genetic modifications that confer long-term ... More