Iodine, 0.1N (0.05M) standard solution
Iodine, 0.1N (0.05M) standard solution
Iodine, 0.1N (0.05M) standard solution
Acros Organics

Iodine, 0.1N (0.05M) standard solution

CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 124220010
HSN 28012000
Price (INR)
11,102.00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Dark brown
Titration with Na2S2O30.0998 to 0.1002 N (20°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solution
RUO – Research Use Only