Hydrochloric acid, pure, fuming, 37% solution in water
Hydrochloric acid, pure, fuming, 37% solution in water
Hydrochloric acid, pure, fuming, 37% solution in water
Acros Organics

Hydrochloric acid, pure, fuming, 37% solution in water

Hydrochloric acid, 35.0 to 39.0%, ClH, CAS Number-7647-01-0, 7732-18-5, acide chlorhydrique, anhydrous hydrochloric acid, chloorwaterstof, chlorohydric acid, chlorwasserstoff, hydrochloric acid, hydrogen chloride, hydrogen chloride hcl, muriatic acid, spirits of salt, 10L, 57.0 deg.C, CHEBI:17883 | CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantityPackaging
124620010
also known as 12462
1 LGlass Bottle
Catalog number 124620010
also known as 12462
HSN 28061000
Price (INR)
4,734.00
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
4,734.00
Each
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Free chlorine=<10 ppm (Cl)
Sulfate (SO4)=<100 ppm
Color scale=<10 APHA
Lead (Pb)=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only