Fetal Bovine Serum, qualified, heat inactivated, Canada
Gibco™

Fetal Bovine Serum, qualified, heat inactivated, Canada

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust ofRead more
Have Questions?
Catalog NumberQuantity
12484010100 mL
Catalog number 12484010
Price (INR)
-
Quantity:
100 mL

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Sera Category: Value (Standard)
  • Origin: Canada
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
Purity or Quality GradeResearch Grade, Qualified
Shipping ConditionFrozen
SpeciesCattle/Bovine
AgeFetal
Country of OriginCanada
FormatBottle
Product TypeFetal Bovine Serum
Quantity100 mL
Serum TreatmentHeat-inactivated
Unit SizeEach
Contents & Storage
Storage conditions: ≤-10°C
Shipping conditions: Frozen

Frequently asked questions (FAQs)

What are the benefits and disadvantages of using heat-inactivated FBS in cell culture?

  1. Heating inactivates complement. Active complement can participate in cytolytic events, contract smooth muscle, release histamine from mast cells and platelets, and activate lymphocytic and macrophage cells. Applications where heat-inactivated serum is recommended include immunological studies and culturing of embryonic stem cells (ESCs), insect cells, and smooth muscle cells.
  2. Heat inactivation helps to achieve bottle-to-bottle and lot-to-lot stability by neutralizing many factors that can vary largely from lot to lot.
  3. There aren't necessarily disadvantages to heat inactivation of FBS, but there is some evidence that suggests there may be no added benefit to it unless you are carrying out immune studies.

Note: Heat inactivation is performed in a 56 degrees C water bath for 30 min with swirling every 10 min or so for heat distribution and to lower the degree of protein aggregation/flocculant precipitation. Note: If the time or temperature is exceeded, the serum may thicken to a gel. If this occurs, the serum is no longer usable. Unnecessary heat inactivation can take up time and potentially lead to wasted reagents if a mistake is made during the protocol1.

1. Pellerin, et al., Bioengineering, published in 2021.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

When should I use heat-inactivated serum?

Heat-inactivation of serum inactivates the “complements” in the serum. In general, it is not necessary to use heat-inactivated serum to culture most cell types. We recommend using heat-inactivated serum when working with immune type cells or for immunological applications.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I would like to order heat-inactivated FBS. What conditions do you use for heat-inactivation of FBS during the manufacturing process?

Serum is heat inactivated by heating at 56 degrees C in a water bath for 30 min, swirling the bottle approximately every 10 minutes.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the difference between "qualified" versus "certified" purity grade FBS?

The main difference is in the Quality Control (QC) test specifications. “Qualified” and “Certified” FBS go through the same QC tests but “Certified” FBS has additional QC tests as well as more stringent QC test specifications for endotoxin and hemoglobin levels. The additional QC tests in “Certified” FBS include biochemical and hormonal profiling.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

If my FBS arrives partially thawed, can I still use it?

Our studies have shown that short-term storage of thawed FBS at 4 degrees C for up to 28 days causes no decrease in growth or viability performance. FBS stored at 4 degrees C for longer periods of time should be evaluated on a case-by-case basis should this storage condition be necessary.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.