GD-MS Consumable Kits
Actual product may vary
Thermo Scientific™
GD-MS Consumable Kits
Optimize your Thermo Scientific™ ELEMENT™ GD GD-MS instrument to your application and analyte of interest with these handy consumable kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1249200Basic Consumables Kit
1227610Element Consumable Kits
1249190High Purity Consumables Kit
Catalog number 1249200
Price (INR)
-
Type:
Basic Consumables Kit
Ensure you have everything needed to optimize your Thermo Scientific™ ELEMENT™ GD GD-MS instrument to your chosen application and analyte of interest with these handy Thermo Scientific™ GD-MS Consumable Kits. Consumable Kits are available in either graphite or steel, according to the detection limits and cleaning procedures required for your analytical task.
Select Quality Consumables for Improved Performance
These convenient and comprehensive kits are tailored for the ELEMENT GD GD-MS analyzer, to fully optimize the system for your analyte of interest
  • High purity consumables kit, with graphite parts, includes cones for the magnetic holder
  • Basic consumables kit, with steel parts for samples of medium purity, includes cones for the magnetic holder
Specifications
For Use With (Equipment)ELEMENT GD GD-MS Instrument
TypeBasic Consumables Kit
Unit SizeEach