Linalool, 97%
Linalool, 97%
Linalool, 97%
Acros Organics

Linalool, 97%

CAS: 78-70-6 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 125150025
also known as 12515
HSN 29052230
Price (INR)
51,462.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
51,462.00
Each
Chemical Identifiers
CAS78-70-6
IUPAC Name3,7-dimethylocta-1,6-dien-3-ol
Molecular FormulaC10H18O
InChI KeyCDOSHBSSFJOMGT-UHFFFAOYNA-N
SMILESCC(C)=CCCC(C)(O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Specific gravity(20/4°C) 0.859 to 0.864
GC>=96.0 %
Specific optical rotation-2° to +2° (20°C, 589 nm) (neat)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only