DL-Menthol, 99%
DL-Menthol, 99%
DL-Menthol, 99%
Acros Organics

DL-Menthol, 99%

CAS: 89-78-1 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 125390050
also known as 12539
HSN 29061100
Price (INR)
4,166.00
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,166.00
Each
Chemical Identifiers
CAS89-78-1
IUPAC Name(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
Molecular FormulaC10H20O
InChI KeyNOOLISFMXDJSKH-KXUCPTDWSA-N
SMILESCC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline solid
Specific optical rotation-2° to +2° (20°C, 589 nm) (c=10, EtOH)
Appearance (Color)Colorless to white
Infrared spectrumConforms
Melting point32°C to 36°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only