L(-)-Menthol, 99.5%
L(-)-Menthol, 99.5%
L(-)-Menthol, 99.5%
Acros Organics

L(-)-Menthol, 99.5%

CAS: 2216-51-5 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 125405000
HSN 29061100
Price (INR)
18,511.00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2216-51-5
IUPAC Name(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
Molecular FormulaC10H20O
InChI KeyNOOLISFMXDJSKH-KXUCPTDWSA-N
SMILESCC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or crystalline needles
Infrared spectrumConforms
GC>=99.4 %
Specific optical rotation-48° to -51° (20°C, 589 nm) (c=10, EtOH)
Appearance (Color)Colorless to white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only