Anode Caps
Actual product may vary
Thermo Scientific™
Anode Caps
This range of top quality Thermo Scientific™ Anode Caps for the Thermo Scientific™ ELEMENT GD™ GD-MS analyzer, ensures consistent, reliable performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
1259210Uncoated Anode Cap 4mm orifice
1187441Anode Caps Stainless Steel
1212710Anode Cap Removal Tool
1212840Anode Cap PyC Coated
1216160Anode Cap Tantalum
1248160Uncoated Anode Cap
Catalog number 1259210
Price (INR)
-
Type:
Uncoated Anode Cap 4mm orifice
This range of top quality Thermo Scientific™ Anode Caps for the Thermo Scientific™ ELEMENT GD™ GD-MS analyzer ensures consistent, reliable performance.
Stainless steel Anode Caps are disposable items, to avoid any carry-over from sample to sample. Graphite parts are designed to be cleaned wet chemically using mineral acids. With the handy removal tool, replacing the Anode Caps is a quick and simple process.
Select Consumables to Match your Analysis Needs
Anode Caps are available in the following materials and order quantities:
  • Stainless steel; general purpose cap, designed for single use to avoid memory effects (50 pieces)
  • Graphite uncoated; general purpose for extra-low detection limits. Can be acid cleaned with every mineral acid, including hydrofluoric acid (singles)
  • Graphite PyC coated; for extra-low detection limits, especially for ppb level analysis of Boron and Phosphorous (20 pieces)
  • Graphite uncoated, with 4mm orifice for small embedded samples (singles)
  • Tantalum; available on request
  • Tool for removing anode caps
Specifications
For Use With (Equipment)ELEMENT GD GD-MS Instrument
TypeUncoated Anode Cap 4mm orifice
Unit SizeEach