Hollow Ball Kit
Thermo Scientific™
Hollow Ball Kit
The Hollow Ball Kit has been discontinued, and is no longer available.
Have Questions?
Catalog NumberQuantityIncludes
126000000020100 Ea.1.5 in. dia. balls
Catalog number 126000000020
Price (INR)
-
Quantity:
100 Ea.
Includes:
1.5 in. dia. balls
The Hollow Ball Kit has been discontinued, and is no longer available.
Insulate your bath reservoir from temperature losses while allowing immersion of a variety of vessels such as flasks or test tubes with the Thermo Scientific™ Hollow Ball Kit.
Includes:
100 1.5in. diameter balls
Specifications
Includes1.5 in. dia. balls
Quantity100 Ea.
Unit SizeEach