Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
TrypLE™ Express Enzyme (1X), no phenol red
Gibco™

TrypLE™ Express Enzyme (1X), no phenol red

TrypLE Express is an animal origin-free, recombinant enzyme used for dissociating a wide range of adherent mammalian cells, including CHO,Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1260403920 x 100mL
12604013100 mL
12604021500 mL
126040545 L
Catalog number 12604039
Price (INR)
51,516.00
Each
Quantity:
20 x 100mL
Customize this product
Price (INR)
51,516.00
Each
TrypLE Express is an animal origin-free, recombinant enzyme used for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. TrypLE Express cleaves peptide bonds on the C-terminal sides of lysine and arginine, and is a direct replacement for trypsin. Its exceptional purity increases specificity and reduces damage to cells that can be caused by other enzymes present in some trypsin extracts. We offer a variety of TrypLE formulations for a range of cell culture applications. Compared to trypsin and other dissociation reagents, TrypLE Express is:

• Gentle on cells
• Room temperature-stable
• Easy to use
• Animal origin-free (AOF)
This TrypLE Express is modified as follows:
WithWithout
• EDTA• Phenol Red

The formulation of TrypLE solutions is available, although we keep the enzyme concentration confidential.

Gentle on cells
TrypLE Express' exceptional purity increases specificity due to the action of a single enzyme. This reduces damage caused by cleavage from multiple enzymes in trypsin and other extracts.

Room temperature-stable
TrypLE Express remains stable for 24 months at room temperature or 2–8°C, making storage and handling easy and convenient.

Easy to use
TrypLE Express can be directly substituted for trypsin in existing protocols. In addition, dilution alone inactivates TrypLE Express, avoiding the need for trypsin inhibitors, such as FBS.

Animal origin-free (AOF)
Unlike porcine trypsin, TrypLE Express is free of animal-derived components.

cGMP manufacturing and quality system
For supply chain continuity, we manufacture Gibco TrypLE Express Enzyme (1X), no phenol red at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
ChelatorsEDTA
Concentration1 X
Culture TypeAdherent Cell Culture
OsmolalityCheck and Record
Product LineGibco
Quantity20 x 100mL
RemarksRoom Temperature Stable
Shelf Life24 Months
Shipping ConditionRoom Temperature
SourceAnimal origin-free
Tested ForIn vitro Bioassay
ClassificationAnimal Origin-free
FormLiquid
Product TypeCell Culture Dissociation Reagent
SterilitySterile-filtered
Without AdditivesNo Phenol Red
pH6 to 8
Unit SizeEach
Contents & Storage
Storage conditions: 15°C to 30°C, protect from light
Alternate storage conditions: 2°C to 8°C or -5°C to -20°C
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture

Frequently asked questions (FAQs)

What enzymes do you recommend for passaging mesenchymal stem cells (MSC)?

We recommend using TrypLE Express Enzyme or TrypLE Select Enzyme for passaging mesenchymal stem cells (MSCs). The usage of trypsin will result in lower cell viability upon passage. TrypLE Express and TrypLE Select are defined, animal origin-free, and stable at both room and refrigerated temperatures.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Does Diploid Growth Serum-Reduced Medium (SRM) contain L-glutamine?

The basal Diploid Growth Serum-Reduced Medium already contains 6 mM L-glutamine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do I have to adjust my infection parameters when using Diploid Production Serum-Free Medium (SFM)?

We recommend evaluating performance with Diploid Production Serum-Free Medium using current conditions, however, multiplicity of infection, time of infection, and time of harvest may be different than with conventional media. Closely monitor cells for cytopathic effect and evaluate viral titers at multiple time points.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can Diploid Growth Serum-Reduced Medium (SRM) be supplemented with different types of serum?

Diploid Growth Serum-Reduced Medium can be supplemented with Fetal Bovine Serum, Newborn Calf Serum, Donor Bovine Serum with Iron, or Bovine Serum.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Which cell lines can I culture in Diploid Growth Serum-Reduced Medium (SRM)?

Diploid Growth Serum-Reduced Medium has been evaluated with fibroblast cell lines, including MRC-5, WI-38, IMR-90, BS-2, and CEF cells.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (4)

Citations & References
Abstract
Isolation and cultivation of human keratinocytes from skin or plucked hair for the generation of induced pluripotent stem cells.
Authors:Aasen T, Izpisúa Belmonte JC,
Journal:Nat Protoc
PubMed ID:20134422
'The ease of generating induced pluripotent stem (iPS) cells, and possibly their properties after reprogramming, depends on the origin of the somatic cell starting population. Reprogramming of keratinocytes is both faster and more efficient compared with fibroblasts, although more care is required when isolating, culturing and infecting these cells. In ... More
VCAM-1 expression in adult hematopoietic and nonhematopoietic cells is controlled by tissue-inductive signals and reflects their developmental origin.
Authors:Ulyanova T, Scott LM, Priestley GV, Jiang Y, Nakamoto B, Koni PA, Papayannopoulou T,
Journal:Blood
PubMed ID:15769895
Although expression of vascular cell adhesion molecule 1 (VCAM-1) in endothelial cells and its functional implications have been previously appreciated, VCAM-1 expression in other than endothelial cells, especially hematopoietic cells, has been recently recognized and has not been explored in detail. Using normal mice and mice with a conditional ablation ... More
A rapid, cost-effective method for counting human embryonic stem cell numbers as clumps.
Authors:Prowse A, Wolvetang E, Gray P,
Journal:Biotechniques
PubMed ID:19594444
Enumeration of human embryonic stem cell (hESC) numbers through single cell digestion can be time consuming especially in high-throughput or multi-factorial analysis containing 50+ samples. We have developed a reproducible, cost-effective method of counting hESCs in clumps circumventing the need to manually dissociate each sample to single cells. The method ... More
Human Synovial MSC Good Manufacturing Practices for Articular Cartilage Regeneration.
Authors:Fernandes TL, Kimura HA, Pinheiro CCG, Shimomura K, Nakamura N, Ferreira JRM, Gomoll AH, Hernandez AJ, Bueno DF,
Journal:Tissue Eng Part C Methods
PubMed ID:30412046
cartilage restoration is a desperately needed bridge for patients with symptomatic cartilage lesions. Chondral lesion is a pathology with high prevalence, reaching as much as 63% of general population and 36% among athletes. Despite Autologous Chondrocyte Implantation (ACI) versatility, it still fails to fully reproduce hyaline articular cartilage characteristics. Mesenchymal ... More