Methyl benzoylformate, 99%
Methyl benzoylformate, 99%
Methyl benzoylformate, 99%
Acros Organics

Methyl benzoylformate, 99%

CAS: 15206-55-0 | C9H8O3 | 164.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 126370250
also known as 12637
HSN 29151300
Price (INR)
9,776.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,776.00
Each
Chemical Identifiers
CAS15206-55-0
IUPAC Namemethyl 2-oxo-2-phenylacetate
Molecular FormulaC9H8O3
InChI KeyYLHXLHGIAMFFBU-UHFFFAOYSA-N
SMILESCOC(=O)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear colorless to yellow
Refractive index1.5250 to 1.5280 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only