Product Image
Gibco™

AlgiMatrix™ 96-Well, 3D Culture System, Flat-Bottom Microplate

Color: Clear; Sterility: Sterile; Lid: With Lid
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityNo. of Wells
126840315 x 96 well plates96
126840151 x 96 well plate96
Catalog number 12684031
Price (INR)
-
Quantity:
5 x 96 well plates
No. of Wells:
96
AlgiMatrix™ 3D Culture System is the first user-friendly non-animal-derived bioscaffold for three-dimensional (3D) cell culture. AlgiMatrix™ 3D Culture System creates a cell culture model system that more closely reflects normal cell morphology and behavior, to meet researchers' requirements for 3D cell culture in fields such as toxicology, drug development, cancer and stem cell research, development and morphogenesis, tissue and organ engineering, heart disease, diabetes, and Alzheimers disease.

AlgiMatrix™ 3D Culture System is supplied in a 96-well format that fits well into the workflow of both lower- and higher-throughput users. Scientists can inoculate cells directly into the sterile microtiter plates preloaded with lyophilized alginate sponge, with no pretreatments or preparations required. The combination of convenience, high capacity, and microscopic visualization makes AlgiMatrix™ 3D Culture System an ideal solution for broad basic research and drug discovery applications.

Applications:
AlgiMatrix™ 3D Culture System is suitable for many cell-based screening, drug discovery, and human cell therapy procedures, including the Multicellular Tumor Spheroid Assays (MCTS),1 hepatocyte 2-4 and cardiomyocyte organogenesis studies,5 co-culture studies,6 high-throughput drug screening assays,1 and embryonic stem cell 3D differentiation.7 Each lot is performance and quality tested in a cell-based toxicity assay.

Storage:
Room temperature.

References:
1. Kunz-Schughart, L., et al., (2004) J Biomolec Screening 9, 273.
2. Dvir-Ginzberg, M., et al. (2003) Tissue Engineering 9, 757.
3. Elkayam, T., et al. (2006) Tissue Engineering 12, 1357.
4. Kedem, A., et al. (2005) Tissue Engineering 11, 715.
5. Dar, A., et al. (2002) Biotechnology & Bioengineering 80, 305.
6. Kim, J. (2005) Seminars in Cancer Biology 15, 365.
7. Gerecht-Nir, S., et al. (2004) Biotechnology & Bioengineering 88, 313.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
LidWith Lid
No. per Pack5/Pk.
Packaging5/Pk.
Product TypeMicroplate
Purity or Quality GradeCell Culture Grade
Quantity5 x 96 well plates
Shipping ConditionRoom Temperature
ColorClear
No. of Wells96
Product LineAlgimatrix
SterilitySterile
Surface TreatmentAlgiMatrix 3D Culture System
Well ShapeFlat Bottom (F-well)
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

How stable is AlgiMatrix Matrix at room temperature without degradation?

AlgiMatrix Matrix is stable for 12 months unopened. Once opened, the entire plate needs to be used.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Why can Gibco Geltrex Matrix be used for adhesion-related research, as opposed to Gibco AlgiMatrix 3D Culture System which cannot?

Geltrex Matrix is a basement membrane extract that contains collagen IV, laminin, and other matrix proteins and growth factors that are required for cell adhesion. AlgiMatrix Matrix contains only alginate which does not facilitate cell adhesion. There is nothing in the alginate sponges for the cells to interact with.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can other cell viability reagents be used with AlgiMatrix Matrix?

Other than the Invitrogen alamarBlue Cell Viability Reagent, AlgiMatrix Matrix is also compatible with Invitrogen PrestoBlue Cell Viability Reagent and Invitrogen CyQuant Direct Cell Proliferation Assay. The protocol is almost the same as the 2D cell culture, but it needs the blank AlgiMatrix Matrix as a blank control.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How important is calcium in the AlgiMatrix firming and dissolving buffers?

The firming buffer works by adding calcium to the sponge, making it stiffer. The dissolving buffer works by removing calcium from the sponge, making it softer.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Is there any assay variability when using AlgiMatrix Matrix?

3D cell culture with AlgiMatrix Matrix approaches a more physiological state in terms of cell growth and behavior, but along with that comes the higher variability typically associated with more complex systems. You may need to do more replicates or more controls.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.