Product Image
Gibco™
AlgiMatrix™ 24-Well, 3D Culture System, Flat-Bottom Microplate
The Algimatrix™ 3D Culture System provides a three-dimensional matrix that has the potential to create cell culture conditions that moreRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityNo. of Wells
126840494 x 24 well plates24
126840231 x 24 well plate24
Catalog number 12684049
Price (INR)
-
Quantity:
4 x 24 well plates
No. of Wells:
24
The Algimatrix™ 3D Culture System provides a three-dimensional matrix that has the potential to create cell culture conditions that more closely reflect normal cell morphology and behavior. It has relevance for both academic and industrial researchers who want to culture hepatocytes and other spheriod forming cells. Algimatrix™ alginate sponge is provided in a 24 well format that fits well into the workflow of both lower and higher-throughput users. The larger well size enables growth of more cells for downstream applications such as transfection.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
LidWith Lid
Packaging4 Pack
Product TypeMicroplate
Purity or Quality GradeCell Culture Grade
Quantity4 x 24 well plates
Shipping ConditionRoom Temperature
ColorClear
No. of Wells24
Product LineAlgimatrix
SterilitySterile
Surface TreatmentAlgiMatrix 3D Culture System
Well ShapeFlat Bottom (F-well)
Unit SizeEach
Contents & Storage
Higher-fidelity, animal-free scaffold for 3D cell culture that mimics in vivo conditions. More consistent results than animal-derived matrices. Ready to use, stored at room temperature and stable up to 18 months. Protocols available.
Frequently asked questions (FAQs)
How stable is AlgiMatrix Matrix at room temperature without degradation?
AlgiMatrix Matrix is stable for 12 months unopened. Once opened, the entire plate needs to be used.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Why can Gibco Geltrex Matrix be used for adhesion-related research, as opposed to Gibco AlgiMatrix 3D Culture System which cannot?
Geltrex Matrix is a basement membrane extract that contains collagen IV, laminin, and other matrix proteins and growth factors that are required for cell adhesion. AlgiMatrix Matrix contains only alginate which does not facilitate cell adhesion. There is nothing in the alginate sponges for the cells to interact with.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can other cell viability reagents be used with AlgiMatrix Matrix?
Other than the Invitrogen alamarBlue Cell Viability Reagent, AlgiMatrix Matrix is also compatible with Invitrogen PrestoBlue Cell Viability Reagent and Invitrogen CyQuant Direct Cell Proliferation Assay. The protocol is almost the same as the 2D cell culture, but it needs the blank AlgiMatrix Matrix as a blank control.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How important is calcium in the AlgiMatrix firming and dissolving buffers?
The firming buffer works by adding calcium to the sponge, making it stiffer. The dissolving buffer works by removing calcium from the sponge, making it softer.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is there any assay variability when using AlgiMatrix Matrix?
3D cell culture with AlgiMatrix Matrix approaches a more physiological state in terms of cell growth and behavior, but along with that comes the higher variability typically associated with more complex systems. You may need to do more replicates or more controls.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.