2-Methylheptane, 99%
2-Methylheptane, 99%
2-Methylheptane, 99%
Acros Organics

2-Methylheptane, 99%

CAS: 592-27-8 | C8H18 | 114.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 126880010
also known as 12688
HSN 29011000
Price (INR)
4,616.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,616.00
Each
Chemical Identifiers
CAS592-27-8
IUPAC Name2-methylheptane
Molecular FormulaC8H18
InChI KeyJVSWJIKNEAIKJW-UHFFFAOYSA-N
SMILESCCCCCC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.3930 to 1.3970 (20°C, 589 nm)
GC>=98.5 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only