Nicotinamide, 99%
Nicotinamide, 99%
Nicotinamide, 99%
Acros Organics

Nicotinamide, 99%

CAS: 98-92-0 | C6H6N2O | 122.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 128275000
HSN 29362910
Price (INR)
8,735.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS98-92-0
IUPAC Namepyridine-3-carboxamide
Molecular FormulaC6H6N2O
InChI KeyDFPAKSUCGFBDDF-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Loss on drying=<0.5 % (4 h) (silica gel)
pH6.0 to 7.5 (5 % in water at 20°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only