Nicotinic acid, 99.5%
Nicotinic acid, 99.5%
Nicotinic acid, 99.5%
Acros Organics

Nicotinic acid, 99.5%

CAS: 59-67-6 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 128290025
also known as 12829
HSN 29362950
Price (INR)
34,679.00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS59-67-6
IUPAC Namepyridine-3-carboxylic acid
Molecular FormulaC6H5NO2
InChI KeyPVNIIMVLHYAWGP-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with NaOH>=99.4 %
Sulfated ash=<0.1 %
Loss on drying=<1 % (105°C, 1 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only