n-Octylamine, 99+%
n-Octylamine, 99+%
n-Octylamine, 99+%
Acros Organics

n-Octylamine, 99+%

CAS: 111-86-4 | C8H19N | 129.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
500 g
Packaging:
Glass bottle
Catalog number 129491000
HSN 29211990
Price (INR)
5,018.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,018.00
Each
Chemical Identifiers
CAS111-86-4
IUPAC Nameoctan-1-amine
Molecular FormulaC8H19N
InChI KeyIOQPZZOEVPZRBK-UHFFFAOYSA-N
SMILESCCCCCCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Refractive index1.4280 to 1.4300 (20°C, 589 nm)
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only