Oleylamine, approximate C18-content 80-90%
Oleylamine, approximate C18-content 80-90%
Oleylamine, approximate C18-content 80-90%
Acros Organics

Oleylamine, approximate C18-content 80-90%

CAS: 112-90-3 | C18H37N | 267.50 g/mol
Have Questions?
Catalog NumberQuantity
129541000
also known as 12954
100 mL
Catalog number 129541000
also known as 12954
HSN 29211990
Price (INR)
6,226.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (INR)
6,226.00
Each
Chemical Identifiers
CAS112-90-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Appearance (Form)Liquid after melting
Infrared spectrumConforms
Titration with HClO4>=96.0 % (Primary amine)
Refractive index1.4585 to 1.4625 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only