Protect the quality of your products or laboratory samples in cleanroom environments by using particle-certified glass containers.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Product Type
Color
No. per Case
131-08CLP
also known as 131-08C/LP
250 mL
Sample Jar
Clear
24
114-250ALP
250 mL
Boston Round Bottle
Amber
12
2 Options
Catalog number 131-08CLP
also known as 131-08C/LP
Price (INR)
-
Capacity (Metric):
250 mL
Product Type:
Sample Jar
Color:
Clear
No. per Case:
24
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
Class 100/10 glass
Certificate of Analysis is included with each shipment
Available in clear or amber
Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
Individual bottles double bagged to maintain integrity until point of use
4 L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
Low particle applications
Specifications
Capacity8 oz., 250 mL
Closure MaterialPolypropylene
Closure Size70-400 mm
DescriptionClear Glass Wide Mouth Jar with Polypropylene Closure, 250 mL