1,2-Phthalic dicarboxaldehyde, 98+%
1,2-Phthalic dicarboxaldehyde, 98+%
1,2-Phthalic dicarboxaldehyde, 98+%
Acros Organics

1,2-Phthalic dicarboxaldehyde, 98+%

CAS: 643-79-8 | C8H6O2 | 134.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 131080050
also known as 13108
HSN 29122990
Price (INR)
11,575.00
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
11,575.00
Each
Chemical Identifiers
CAS643-79-8
IUPAC Namebenzene-1,2-dicarbaldehyde
Molecular FormulaC8H6O2
InChI KeyZWLUXSQADUDCSB-UHFFFAOYSA-N
SMILESO=CC1=CC=CC=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or lumps
Melting point54°C to 58°C
GC>=98.0 %
Appearance (Color)Yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only