(1S)-(-)-alpha-Pinene, 98%
(1S)-(-)-alpha-Pinene, 98%
(1S)-(-)-alpha-Pinene, 98%
Acros Organics

(1S)-(-)-alpha-Pinene, 98%

CAS: 7785-26-4 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 131272500
also known as 13127
HSN 29029090
Price (INR)
6,178.00
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
6,178.00
Each
Chemical Identifiers
CAS7785-26-4
IUPAC Name(1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-ene
Molecular FormulaC10H16
InChI KeyGRWFGVWFFZKLTI-IUCAKERBSA-N
SMILESCC1=CC[C@H]2C[C@@H]1C2(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Specific optical rotation-35° to -45° (20°C, 589 nm) (Neat)
GC>=97.5 %
Appearance (Form)Clear liquid
Refractive index1.4640 to 1.4680 (20°C, 589 nm)
Specific gravity(20°C) 0.858 to 0.861
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only