DL-Pipecolinic acid, 99%
DL-Pipecolinic acid, 99%
DL-Pipecolinic acid, 99%
Acros Organics

DL-Pipecolinic acid, 99%

CAS: 535-75-1 | C6H11NO2 | 129.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 131280250
also known as 13128
HSN 29333990
Price (INR)
7,007.00
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
7,007.00
Each
Chemical Identifiers
CAS535-75-1
IUPAC Namepiperidine-2-carboxylic acid
Molecular FormulaC6H11NO2
InChI KeyHXEACLLIILLPRG-UHFFFAOYNA-N
SMILESOC(=O)C1CCCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point268°C to 278°C (Dec)
Titration with HClO4>=98.5 %
Water=<1 % (K.F.)
Appearance (Color)White to light beige
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only