Pyrrole-2-carboxaldehyde, 99%
Pyrrole-2-carboxaldehyde, 99%
Pyrrole-2-carboxaldehyde, 99%
Acros Organics

Pyrrole-2-carboxaldehyde, 99%

CAS: 1003-29-8 | C5H5NO | 95.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 132061000
also known as 13206
HSN 29339900
Price (INR)
45,757.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
45,757.00
Each
Chemical Identifiers
CAS1003-29-8
IUPAC Name1H-pyrrole-2-carbaldehyde
Molecular FormulaC5H5NO
InChI KeyZSKGQVFRTSEPJT-UHFFFAOYSA-N
SMILESO=CC1=CC=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Appearance (Form)Low melting crystalline solid
AppearanceMay darken on storage
Infrared spectrumConforms
Melting point40°C to 46°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only