Autosampler Housing
Thermo Scientific™
Autosampler Housing
Eliminate the risk of contamination from the laboratory environment with this dedicated Thermo Scientific™ Autosampler Housing, designed for our Thermo Scientific™ iCAP™ Q ICP-MS system.
Have Questions?
Catalog NumberTypeFor Use With (Equipment)
1322190Autosampler HousingiCAP Q ICP-MS System
Catalog number 1322190
Price (INR)
-
Type:
Autosampler Housing
For Use With (Equipment):
iCAP Q ICP-MS System
Eliminate the risk of contamination from the laboratory environment with this dedicated Thermo Scientific™ Autosampler Housing, designed especially for the Thermo Scientific™ iCAP™ Q ICP-MS system.
Compatible with the range of autosamplers from CETAC™ and ESI™ to offer maximum choice and flexibility; this autosampler housing provides a clean environment for samples before and during analysis to reduce false positives and save on the costs of analysis re-runs.
Increase Accuracy with the Right Accessories
  • Autosampler housing mounted directly onto the side of the iCAP Q ICP-MS analyzer
  • Compatible with all standard iCAP Q ICP-MS system sample introduction systems
  • Four rack random access autosamplers from ESI or CETAC can be installed inside the autosampler housing
  • Positive ULPA filtered air pressure supplied to the autosampler housing ensures clean sample environment throughout analysis
Specifications
DescriptionAutosampler Housing
For Use With (Equipment)iCAP Q ICP-MS System
TypeAutosampler Housing
Unit SizeEach