Product Image
Thermo Scientific™
Collision Cell Technology Upgrade Kit
Expand your Thermo Scientific™ iCAP™ Qa ICP-MS system capabilities as your laboratory requirements advance with this QCell ion guide in-field upgrade kit.
Have Questions?
Catalog NumberTypeFor Use With (Equipment)
1324700Collision Cell Technology Upgrade KitiCAP Qa ICP-MS System
Catalog number 1324700
Price (INR)
-
Type:
Collision Cell Technology Upgrade Kit
For Use With (Equipment):
iCAP Qa ICP-MS System
Advance your Thermo Scientific™ iCAP™ Qa ICP-MS instrument performance as your laboratory requirements progress.
With this Thermo Scientific™ Collision Cell Technology Upgrade Kit, the Thermo Scientific™ QCell ion guide can be upgraded in the field, to provide full reaction and collision cell capabilities for your iCAP Qa ICP-MS system. Take advantage of the He kinetic energy discrimination (KED) collision cell capabilities, for clearer ICP-MS spectra and better detection power for interfered isotopes in all sample types.
Upgrade Performance In-Field for Maximum Convenience
  • Full KED collision cell and CCT reaction cell capabilities are standard on the iCAP Qc and iCAP Qs ICP-MS models. With this field upgrade kit, the iCAP Qa ICP-MS system can provide the same proprietary QCell performance and your instrument never needs to move off-site.
  • The QCell is an advanced flatapole technology, combining proven He KED interference reduction capability, with unique low mass cut-off ability, for ultra reliable cell-based analyses.
Specifications
DescriptionCollision Cell Technology Upgrade Kit (only applicable for iCAP Qa)
For Use With (Equipment)iCAP Qa ICP-MS System
TypeCollision Cell Technology Upgrade Kit
Unit SizeEach