Product Image
Thermo Scientific™
Precision™ and Lindberg/Blue M™ Oven Shelf Kit
Increase the flexibility of Thermo Scientific™ Precision™ and Lindberg/BlueM™ ovens with these shelf kits.
Have Questions?
Catalog NumberFor Use With (Equipment)
13247S2.5 to 5 cu. ft. Units, Incubator
Catalog number 13247S
Price (INR)
-
For Use With (Equipment):
2.5 to 5 cu. ft. Units, Incubator
Utilize the Thermo Scientific™ Precision™ and Lindberg/BlueM™ Oven Shelf Kit to increase the flexibility of your laboratory oven.
  • Chrome-plated steel, wire-frame style
  • No-tip design
  • With shelf supports
  • Fits oven oven 24 in.W x 23 in.D
Specifications
DescriptionShelf Kit
MaterialChrome Plated
For Use With (Equipment)2.5 to 5 cu. ft. Units, Incubator
TypeShelf Kit
Unit SizeEach