D(-)-Salicin, 99+%
D(-)-Salicin, 99+%
D(-)-Salicin, 99+%
Acros Organics

D(-)-Salicin, 99+%

CAS: 138-52-3 | C13H18O7 | 286.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 132591000
also known as 13259
HSN 29389090
Price (INR)
49,047.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
49,047.00
Each
Chemical Identifiers
CAS138-52-3
IUPAC Name(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
Molecular FormulaC13H18O7
InChI KeyNGFMICBWJRZIBI-UJPOAAIJSA-N
SMILESOC[C@H]1O[C@@H](OC2=CC=CC=C2CO)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.0 %
Water=<0.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only