Semicarbazide hydrochloride, 99+%
Semicarbazide hydrochloride, 99+%
Semicarbazide hydrochloride, 99+%
Acros Organics

Semicarbazide hydrochloride, 99+%

CAS: 563-41-7 | CH6ClN3O | 111.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 132645000
also known as 13264
HSN 29280090
Price (INR)
9,469.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
9,469.00
Each
Chemical Identifiers
CAS563-41-7
IUPAC Namehydrogen aminourea chloride
Molecular FormulaCH6ClN3O
InChI KeyXHQYBDSXTDXSHY-UHFFFAOYSA-N
SMILES[H+].[Cl-].NNC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Solubility(10 % in water) Clear colorless
Appearance (Color)White
Appearance (Form)Crystalline powder and/or chunks
Titration with KIO3>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only