Suberic acid, 99%
Suberic acid, 99%
Suberic acid, 99%
Acros Organics

Suberic acid, 99%

CAS: 505-48-6 | C8H14O4 | 174.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 132815000
also known as 13281
HSN 29171990
Price (INR)
28,879.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
28,879.00
Each
Chemical Identifiers
CAS505-48-6
IUPAC Nameoctanedioic acid
Molecular FormulaC8H14O4
InChI KeyTYFQFVWCELRYAO-UHFFFAOYSA-N
SMILESOC(=O)CCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to cream
Infrared spectrumConforms
Titration with NaOH98.5 to 101.5 %
Melting point141°C to 144°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only