Sulfuric acid, 96%, extra pure, solution in water
Sulfuric acid, 96%, extra pure, solution in water
Sulfuric acid, 96%, extra pure, solution in water
Acros Organics

Sulfuric acid, 96%, extra pure, solution in water

CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Plastic Bottle
Glass Bottle
Catalog number 133610011
HSN 28070010
Price (INR)
4,971.00
Each
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (INR)
4,971.00
Each
Chemical Identifiers
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH95 to 98 %
Arsenic (As)=<0.03 ppm
Appearance (Form)Clear liquid
Chloride (Cl)=<0.5 ppm
Heavy metals (as Pb)=<2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only