Hydrogen chloride, pure, 5 to 6N solution in 2-propanol
Hydrogen chloride, pure, 5 to 6N solution in 2-propanol
Hydrogen chloride, pure, 5 to 6N solution in 2-propanol
Acros Organics

Hydrogen chloride, pure, 5 to 6N solution in 2-propanol

CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
View all
Plastic Bottle
Catalog number 133700025
also known as 13370
HSN 28061000
Price (INR)
36,738.00
Each
Quantity:
2.5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (INR)
36,738.00
Each
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Water=<1.0 %
Appearance (Color)Clear colorless to light yellow
Titration with NaOH5 to 6 N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only