Sodium sulfide hydrate, 60-64%, extra pure, flakes, Thermo Scientific Chemicals
Sodium sulfide hydrate, 60-64%, extra pure, flakes, Thermo Scientific Chemicals
Sodium sulfide hydrate, 60-64%, extra pure, flakes, Thermo Scientific Chemicals
Acros Organics

Sodium sulfide hydrate, 60-64%, extra pure, flakes, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 133720010
HSN 28301000
Price (INR)
5,752.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
5,752.00
Each
Chemical Identifiers
CAS27610-45-3
IUPAC Namedisodium sulfanediide
Molecular FormulaNa2S
InChI KeyGRVFOGOEDUUMBP-UHFFFAOYSA-N
SMILES[Na+].[Na+].[S--]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to pink
Titration Iodimetric60 to 64 % (Na2S)
Sodium carbonate (Na2CO3)=<1.2 %
Iron (Fe)=<10 ppm
Appearance (Form)Flakes
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only