4-Iodotoluene, 98%
4-Iodotoluene, 98%
4-Iodotoluene, 98%
Acros Organics

4-Iodotoluene, 98%

CAS: 624-31-7 | C7H7I | 218.037 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
100 g
Packaging:
Glass Bottle
Catalog number 133940100
HSN 29039990
Price (INR)
7,314.00
Each
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
7,314.00
Each
Chemical Identifiers
CAS624-31-7
IUPAC Name1-iodo-4-methylbenzene
Molecular FormulaC7H7I
InChI KeyUDHAWRUAECEBHC-UHFFFAOYSA-N
SMILESCC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Low melting crystalline mass
Infrared spectrumConforms
Melting point33°C to 36°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only