2-Acetyl-6-methoxynaphthalene, 98%
2-Acetyl-6-methoxynaphthalene, 98%
2-Acetyl-6-methoxynaphthalene, 98%
2-Acetyl-6-methoxynaphthalene, 98%
Acros Organics

2-Acetyl-6-methoxynaphthalene, 98%

CAS: 3900-45-6 | C13H12O2 | 200.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number 134780100
also known as 13478
HSN 29093090
Price (INR)
22,370.00
Each
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
22,370.00
Each
Add to cart
Chemical Identifiers
CAS3900-45-6
IUPAC Name1-(6-methoxynaphthalen-2-yl)ethan-1-one
Molecular FormulaC13H12O2
InChI KeyGGWCZBGAIGGTDA-UHFFFAOYSA-N
SMILESCOC1=CC=C2C=C(C=CC2=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point106°C to 109°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only