N,N,N',N'-Tetramethylbenzidine, 97.5%
N,N,N',N'-Tetramethylbenzidine, 97.5%
N,N,N',N'-Tetramethylbenzidine, 97.5%
Acros Organics

N,N,N',N'-Tetramethylbenzidine, 97.5%

CAS: 366-29-0 | C16H20N2 | 240.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 138380050
also known as 13838
HSN 29215990
Price (INR)
13,209.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
13,209.00
Each
Chemical Identifiers
CAS366-29-0
IUPAC NameN4,N4,N4',N4'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular FormulaC16H20N2
InChI KeyYRNWIFYIFSBPAU-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Infrared spectrumConforms
Titration with HClO4>=96.5 %
Appearance (Form)Powder
Melting point192°C to 196°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only